Assay Details
| Gene Symbol: ACYP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320587.1 | NP_001307516.1 | 2 - 3 | 319 | 91 | |
| XM_017005409.1 | XP_016860898.1 | 2 - 3 | 584 | 91 | ||
| GenBank mRNA | BG741807.1 | - | 2 - 3 | 313 | 91 | |
More Information
Set Membership: |
|

ACYP2
Genomic Map