Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321541.1 | NP_001308470.1 | 2 - 3 | 959 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DEGS1| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321541.1 | NP_001308470.1 | 2 - 3 | 959 | 73 | |
Set Membership: |
|