Assay Details
| Gene Symbol: RPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013.3 | NP_001004.2 | 33 | 69 | ||
| NM_001321705.1 | NP_001308634.1 | 1 - 2 | 33 | 69 | ||
| NM_001321706.1 | NP_001308635.1 | 1 - 2 | 33 | 69 | ||
| NR_135763.1 | 1 - 2 | 33 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001013.3, NM_001321705.1, NM_001321706.1 transcripts.
Set Membership: |
|

RPS9
Genomic Map