Assay Details
Gene Symbol: ASAH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321957.1 | NP_001308886.1 | 1 - 2 | 294 | 62 | |
NM_001321958.1 | NP_001308887.1 | 1 - 2 | 294 | 62 | ||
NR_135904.1 | 1 - 2 | 294 | 62 | |||
GenBank mRNA | AK294369.1 | - | 1 - 2 | 46 | 62 | |
BI459539.1 | - | 1 - 2 | 299 | 62 | ||
CN428611.1 | - | 1 - 2 | 36 | 62 | ||
DC329838.1 | - | 1 - 2 | 54 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK294369.1, NM_001321957.1, NM_001321958.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
