Assay Details
| Gene Symbol: ARPP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017021871.1 | XP_016877360.1 | 1 - 2 | 540 | 89 | |
| GenBank mRNA | AK123616.1 | - | 1 - 2 | 540 | 89 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017021871.1 transcripts.
Set Membership: |
|

ARPP19
Genomic Map