Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182614.2 | NP_872420.1 | 6 - 7 | 532 | 117 | |
| XM_005272413.3 | XP_005272470.1 | 7 - 8 | 651 | 117 | ||
| XM_005272415.2 | XP_005272472.1 | 7 - 8 | 633 | 117 | ||
| XM_006719986.2 | XP_006720049.1 | 4 - 5 | 327 | 117 | ||
| XM_017020558.1 | XP_016876047.1 | 8 - 9 | 723 | 117 | ||
| XM_017020559.1 | XP_016876048.1 | 9 - 10 | 1932 | 117 | ||
| XM_017020560.1 | XP_016876049.1 | 5 - 6 | 495 | 117 | ||
| GenBank mRNA | BC018995.2 | - | 6 - 7 | 532 | 4300679 | 117 |
More Information
Set Membership: |
|

TMEM255B
Genomic Map