Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000277.1 | NP_000268.1 | 5 - 6 | 982 | 98 | |
| XM_017019370.1 | XP_016874859.1 | 5 - 6 | 625 | 98 | ||
| GenBank mRNA | AK298419.1 | - | 5 - 6 | 606 | 98 | |
| AK313383.1 | - | 6 - 7 | 607 | 98 | ||
| K03020.1 | - | 5 - 6 | 732 | 98 | ||
| L47726.1 | - | 5 - 6 | 982 | 98 | ||
| U49897.1 | - | 5 - 6 | 982 | 98 | ||
More Information
Set Membership: |
|

PAH
Genomic Map