Assay Details
| Gene Symbol: PAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000277.1 | NP_000268.1 | 9 - 10 | 1447 | 91 | |
| GenBank mRNA | AK313383.1 | - | 10 - 11 | 1072 | 91 | |
| BC026251.1 | - | 9 - 10 | 1128 | 4767736 | 91 | |
| K03020.1 | - | 9 - 10 | 1197 | 91 | ||
| L47726.1 | - | 9 - 10 | 1447 | 91 | ||
| U49897.1 | - | 9 - 10 | 1447 | 91 | ||
More Information
Set Membership: |
|

PAH
Genomic Map