Assay Details
Gene Symbol: PTGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000954.5 | NP_000945.3 | 3 - 4 | 400 | 65 | |
GenBank mRNA | AK075333.1 | - | 3 - 4 | 400 | 65 | |
AK312817.1 | - | 3 - 4 | 387 | 65 | ||
AY026356.1 | - | 3 - 4 | 325 | 65 | ||
BC005939.1 | - | 3 - 4 | 334 | 4294999 | 65 | |
BC041463.1 | - | 5 - 6 | 1608 | 65 | ||
BM686612.1 | - | 3 - 4 | 388 | 65 | ||
BT019921.1 | - | 3 - 4 | 325 | 65 | ||
BT019922.1 | - | 3 - 4 | 325 | 65 | ||
M61900.2 | - | 3 - 4 | 390 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
