Assay Details
| Gene Symbol: RANBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278639.1 | NP_001265568.1 | 5 - 6 | 987 | 117 | |
| NM_002882.3 | NP_002873.1 | 5 - 6 | 735 | 117 | ||
| XM_011530291.2 | XP_011528593.1 | 5 - 6 | 1049 | 117 | ||
| GenBank mRNA | CA489474.1 | - | 5 - 6 | 677 | 117 | |
| CR456556.1 | - | 5 - 6 | 563 | 117 | ||
| X83617.1 | - | 5 - 6 | 611 | 117 | ||
More Information
Set Membership: |
|

RANBP1
Genomic Map