Assay Details
| Gene Symbol: GALR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003857.3 | NP_003848.1 | 1 - 2 | 449 | 104 | |
| XM_011525427.2 | XP_011523729.1 | 3 - 4 | 1606 | 104 | ||
| GenBank mRNA | AF040630.1 | - | 1 - 2 | 478 | 104 | |
| AF080586.1 | - | 1 - 2 | 391 | 104 | ||
| BC069130.1 | - | 1 - 2 | 447 | 7216986 | 104 | |
| BC074914.2 | - | 1 - 2 | 413 | 30915339 | 104 | |
| BC074915.2 | - | 1 - 2 | 413 | 30915558 | 104 | |
| BC109051.1 | - | 1 - 2 | 416 | 40031879 | 104 | |
| BC109052.1 | - | 1 - 2 | 415 | 40031882 | 104 | |
| EF577401.1 | - | 1 - 2 | 366 | 104 | ||
More Information
Set Membership: |
|

GALR2
Genomic Map