Assay Details
| Gene Symbol: CDC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004359.1 | NP_004350.1 | 2 - 3 | 459 | 74 | |
| XM_005259690.3 | XP_005259747.1 | 2 - 3 | 472 | 74 | ||
| XM_006722952.2 | XP_006723015.1 | 2 - 3 | 472 | 74 | ||
| GenBank mRNA | AK291554.1 | - | 2 - 3 | 461 | 74 | |
| BC009850.2 | - | 2 - 3 | 459 | 3942735 | 74 | |
| BC018143.2 | - | 2 - 3 | 462 | 3884179 | 74 | |
| BC023979.1 | - | 2 - 3 | 436 | 4103120 | 74 | |
| BT006659.1 | - | 2 - 3 | 260 | 74 | ||
| L22005.1 | - | 2 - 3 | 446 | 74 | ||
More Information
Set Membership: |
|

CDC34
Genomic Map