Assay Details
| Gene Symbol: SCGB1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006552.1 | NP_006543.1 | 1 - 2 | 106 | 68 | |
| GenBank mRNA | AJ224171.1 | - | 1 - 2 | 106 | 68 | |
| BC062693.1 | - | 1 - 2 | 136 | 30327780 | 68 | |
| BC069170.1 | - | 1 - 2 | 106 | 7216920 | 68 | |
| BC069289.1 | - | 1 - 2 | 106 | 7262693 | 68 | |
More Information
Set Membership: |
|

SCGB1D1
Genomic Map