Assay Details
| Gene Symbol: VPREB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007128.3 | NP_009059.1 | 1 - 2 | 75 | 71 | |
| GenBank mRNA | BC017922.1 | - | 1 - 2 | 67 | 71 | |
| CR456609.1 | - | 1 - 2 | 52 | 71 | ||
More Information
Set Membership: |
|

VPREB1
Genomic Map