Assay Details
| Gene Symbol: NDOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144026.2 | NP_001137498.1 | 11 - 12 | 1550 | 73 | |
| NM_001144027.2 | NP_001137499.1 | 10 - 11 | 1448 | 73 | ||
| NM_001144028.2 | NP_001137500.1 | 11 - 12 | 1529 | 73 | ||
| NM_014434.3 | NP_055249.1 | 11 - 12 | 1550 | 73 | ||
| XM_011518545.1 | XP_011516847.1 | 10 - 11 | 1438 | 73 | ||
| XM_011518547.1 | XP_011516849.1 | 11 - 12 | 1340 | 73 | ||
| XM_011518548.2 | XP_011516850.1 | 10 - 11 | 2740 | 73 | ||
| XM_017014626.1 | XP_016870115.1 | 11 - 12 | 1550 | 73 | ||
| XM_017014627.1 | XP_016870116.1 | 11 - 12 | 1550 | 73 | ||
| GenBank mRNA | AF199509.1 | - | 11 - 12 | 1486 | 73 | |
| AK026089.1 | - | 11 - 12 | 1522 | 73 | ||
| AK074403.1 | - | 10 - 11 | 1409 | 73 | ||
| AK225995.1 | - | 11 - 12 | 1521 | 73 | ||
| AK290026.1 | - | 11 - 12 | 1547 | 73 | ||
| AY077845.1 | - | 11 - 12 | 1468 | 73 | ||
| BC015735.1 | - | 11 - 12 | 1486 | 4869561 | 73 | |
| BC078668.1 | - | 11 - 12 | 1486 | 73 | ||
| BC093782.1 | - | 11 - 12 | 1467 | 7939627 | 73 | |
| BC111943.1 | - | 11 - 12 | 1467 | 8327411 | 73 | |
More Information
Set Membership: |
|

NDOR1
Genomic Map