Assay Details
| Gene Symbol: TYSND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040273.2 | NP_001035363.1 | 1 - 2 | 1320 | 85 | |
| GenBank mRNA | BC030242.1 | - | 1 - 2 | 1236 | 5176098 | 85 |
More Information
Set Membership: |
|

TYSND1
Genomic Map