Assay Details
| Gene Symbol: TAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077504.1 | NP_001070972.1 | 3 - 4 | 234 | 92 | |
| GenBank mRNA | AF515830.1 | - | 3 - 4 | 234 | 92 | |
More Information
Set Membership: |
|

TAC4
Genomic Map