Assay Details
| Gene Symbol: POLR3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016310.4 | NP_057394.3 | 2 - 3 | 247 | 69 | |
| GenBank mRNA | AF051316.1 | - | 2 - 3 | 225 | 69 | |
| AF060223.1 | - | 2 - 3 | 224 | 69 | ||
| BC011932.2 | - | 2 - 3 | 208 | 4303428 | 69 | |
More Information
Set Membership: |
|

POLR3K
Genomic Map