Assay Details
| Gene Symbol: FBXW11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012300.2 | NP_036432.2 | 11 - 12 | 1837 | 74 | |
| NM_033644.2 | NP_387448.2 | 11 - 12 | 1798 | 74 | ||
| NM_033645.2 | NP_387449.2 | 10 - 11 | 1735 | 74 | ||
| XM_005265855.4 | XP_005265912.1 | 12 - 13 | 1887 | 74 | ||
| XM_005265856.2 | XP_005265913.1 | 13 - 14 | 1857 | 74 | ||
| XM_005265857.2 | XP_005265914.1 | 10 - 11 | 1395 | 74 | ||
| XM_005265858.2 | XP_005265915.1 | 12 - 13 | 1899 | 74 | ||
| XM_017009279.1 | XP_016864768.1 | 11 - 12 | 1697 | 74 | ||
| XM_017009280.1 | XP_016864769.1 | 12 - 13 | 1713 | 74 | ||
| XM_017009281.1 | XP_016864770.1 | 12 - 13 | 1781 | 74 | ||
| GenBank mRNA | AB014596.1 | - | 11 - 12 | 1491 | 74 | |
| AB033279.1 | - | 10 - 11 | 1465 | 74 | ||
| AB033280.1 | - | 11 - 12 | 1583 | 74 | ||
| AB033281.1 | - | 11 - 12 | 1605 | 74 | ||
| AF176022.1 | - | 11 - 12 | 1490 | 74 | ||
| AK294956.1 | - | 11 - 12 | 1480 | 74 | ||
| AK314999.1 | - | 10 - 11 | 1364 | 74 | ||
| BC026213.1 | - | 11 - 12 | 1798 | 4419029 | 74 | |
More Information
Set Membership: |
|

FBXW11
Genomic Map