Assay Details
Gene Symbol: C3orf30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152539.2 | NP_689752.2 | 1 - 2 | 1439 | 146 | |
GenBank mRNA | AK057421.1 | - | 1 - 2 | 1439 | 146 | |
BC130475.1 | - | 1 - 2 | 1438 | 40146506 | 146 | |
BC144197.1 | - | 1 - 2 | 1438 | 9052717 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
