Assay Details
Gene Symbol: KLHL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153270.2 | NP_695002.1 | 1 | 2117 | 91 | |
GenBank mRNA | AK092279.1 | - | 1 | 2060 | 91 | |
BC109031.2 | - | 1 | 2029 | 40027951 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
