Assay Details
| Gene Symbol: CD8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172102.3 | NP_742100.1 | 4 - 5 | 590 | 116 | |
| NM_172213.3 | NP_757362.1 | 5 - 6 | 680 | 206 | ||
| GenBank mRNA | BC100911.1 | - | 5 - 6 | 671 | 40003720 | 206 |
| BC100913.1 | - | 5 - 6 | 671 | 40003724 | 206 | |
| BC100914.1 | - | 5 - 6 | 671 | 40003725 | 206 | |
| BQ004743.1 | - | 1 - 2 | 365 | 206 | ||
| X13446.1 | - | 4 - 5 | 581 | 116 | ||
More Information
Set Membership: |
|

CD8B
Genomic Map