Assay Details
| Gene Symbol: QRFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198180.2 | NP_937823.1 | 2 - 3 | 520 | 70 | |
| GenBank mRNA | CV575364.1 | - | 2 - 3 | 520 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_198180.2 transcripts.
Set Membership: |
|

QRFP
Genomic Map