Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198716.1 | NP_942009.1 | 2 - 3 | 1305 | 82 | |
| GenBank mRNA | BC024229.1 | - | 2 - 3 | 1315 | 4660371 | 82 |
| D38300.1 | - | 2 - 3 | 1305 | 82 | ||
| U13214.1 | - | 2 - 3 | 1130 | 82 | ||
| X83861.1 | - | 2 - 3 | 1281 | 82 | ||
More Information
Set Membership: |
|

PTGER3
Genomic Map