Assay Details
| Gene Symbol: PTGER3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198716.1 | NP_942009.1 | 3 - 4 | 1332 | 85 | |
| GenBank mRNA | BC024229.1 | - | 3 - 4 | 1342 | 4660371 | 85 |
| D38300.1 | - | 3 - 4 | 1332 | 85 | ||
| U13214.1 | - | 3 - 4 | 1157 | 85 | ||
| X83861.1 | - | 3 - 4 | 1308 | 85 | ||
More Information
Set Membership: |
|

PTGER3
Genomic Map