Assay Details
Gene Symbol: C16orf92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109659.1 | NP_001103129.1 | 3 - 4 | 317 | 67 | |
NM_001109660.1 | NP_001103130.1 | 3 - 4 | 397 | 67 | ||
GenBank mRNA | AK058133.1 | - | 3 - 4 | 397 | 67 | |
BC101433.1 | - | 3 - 4 | 317 | 40021839 | 67 | |
BC101434.1 | - | 3 - 4 | 317 | 40021838 | 67 | |
BC101435.1 | - | 3 - 4 | 317 | 40021831 | 67 | |
BC101436.1 | - | 3 - 4 | 397 | 40021833 | 67 | |
BC127955.1 | - | 3 - 4 | 397 | 40023544 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
