Assay Details
| Gene Symbol: SPNS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB441165.1 | - | 6 - 7 | 935 | 57 | |
| BC041772.1 | - | 6 - 7 | 1148 | 4814561 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPNS2| Gene Symbol: SPNS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB441165.1 | - | 6 - 7 | 935 | 57 | |
| BC041772.1 | - | 6 - 7 | 1148 | 4814561 | 57 | |
Set Membership: |
|