Assay Details
| Gene Symbol: ZAR1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136571.1 | NP_001130043.1 | 2 - 3 | 774 | 102 | |
| GenBank mRNA | BC146956.1 | - | 2 - 3 | 772 | 9056884 | 102 |
| BC146965.1 | - | 2 - 3 | 774 | 9056893 | 102 | |
More Information
Set Membership: |
|

ZAR1L
Genomic Map