Assay Details
| Gene Symbol: VMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144939.1 | NP_001138411.1 | 2 - 3 | 431 | 80 | |
| GenBank mRNA | CD367473.1 | - | 1 - 2 | 391 | 80 | |
More Information
Set Membership: |
|

VMO1
Genomic Map