Assay Details
| Gene Symbol: YAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190980.2 | NP_001177909.1 | 2 - 3 | 340 | 81 | |
| GenBank mRNA | AK127531.1 | - | 2 - 3 | 220 | 81 | |
More Information
Set Membership: |
|

YAF2
Genomic Map