Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198902.1 | NP_001185831.1 | 1 - 2 | 67 | 76 | |
| GenBank mRNA | BG325066.1 | - | 1 - 2 | 68 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198902.1 transcripts.
Set Membership: |
|

YY1AP1
Genomic Map