Assay Details
Gene Symbol: ATP6V1F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198909.1 | NP_001185838.1 | 1 - 2 | 258 | 93 | |
GenBank mRNA | BU956696.1 | - | 1 - 2 | 157 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
