Assay Details
| Gene Symbol: ATP6V1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198909.1 | NP_001185838.1 | 2 - 3 | 340 | 100 | |
| GenBank mRNA | BU956696.1 | - | 2 - 3 | 239 | 100 | |
More Information
Set Membership: |
|

ATP6V1F
Genomic Map