Assay Details
| Gene Symbol: DCTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199011.1 | NP_001185940.1 | 1 - 2 | 229 | 65 | |
| NM_001199743.1 | NP_001186672.1 | 1 - 2 | 229 | 65 | ||
| NM_032486.3 | NP_115875.1 | 1 - 2 | 229 | 65 | ||
| NR_037573.1 | 1 - 2 | 229 | 65 | |||
| GenBank mRNA | AK027387.1 | - | 1 - 2 | 92 | 65 | |
| AK292843.1 | - | 1 - 2 | 156 | 65 | ||
| AK299631.1 | - | 1 - 2 | 126 | 65 | ||
| BC004191.2 | - | 1 - 2 | 109 | 3505855 | 65 | |
| CD388246.1 | - | 1 - 2 | 148 | 65 | ||
| CR745557.1 | - | 1 - 2 | 66 | 65 | ||
| DA310703.1 | - | 1 - 2 | 229 | 65 | ||
| DB087566.1 | - | 1 - 2 | 85 | 65 | ||
More Information
Set Membership: |
|

DCTN5
Genomic Map