Assay Details
| Gene Symbol: LEMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199052.1 | NP_001185981.1 | 2 - 3 | 218 | 76 | |
| GenBank mRNA | AB084764.1 | - | 2 - 3 | 185 | 76 | |
| AB096682.1 | - | 2 - 3 | 185 | 76 | ||
More Information
Set Membership: |
|

LEMD1
Genomic Map