Assay Details
| Gene Symbol: TCEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204857.1 | NP_001191786.1 | 2 - 3 | 100 | 97 | |
| GenBank mRNA | AL832244.1 | - | 2 - 3 | 101 | 97 | |
| BC093065.1 | - | 2 - 3 | 99 | 30411791 | 97 | |
| BC100028.1 | - | 2 - 3 | 117 | 6062030 | 97 | |
| BG428202.1 | - | 2 - 3 | 100 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC093065.1, BC100028.1, NM_001204857.1 transcripts.
Set Membership: |
|

TCEB1
Genomic Map