Assay Details
| Gene Symbol: TCEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204862.1 | NP_001191791.1 | 3 - 4 | 152 | 102 | |
| GenBank mRNA | CX166168.1 | - | 3 - 4 | 136 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001204862.1 transcripts.
Set Membership: |
|

TCEB1
Genomic Map