Assay Details
| Gene Symbol: PPP5D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205281.1 | NP_001192210.1 | 3 - 4 | 583 | 70 | |
| GenBank mRNA | BM718195.1 | - | 3 - 4 | 315 | 70 | |
More Information
Set Membership: |
|

PPP5D1
Genomic Map