Assay Details
Gene Symbol: OR2Z1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004699.2 | NP_001004699.1 | 1 | 923 | 79 | |
GenBank mRNA | BC137046.1 | - | 1 | 922 | 9021042 | 79 |
BC151149.1 | - | 1 | 923 | 9021043 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
