Assay Details
Gene Symbol: OR8J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005205.2 | NP_001005205.2 | 1 | 822 | 118 | |
GenBank mRNA | BC137066.1 | - | 1 | 9021063 | 118 | |
BC137067.1 | - | 1 | 9021064 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
