Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242369.1 | NP_001229298.1 | 4 - 5 | 529 | 83 | |
| NM_001242370.1 | NP_001229299.1 | 3 - 4 | 403 | 83 | ||
| XM_011518181.2 | XP_011516483.1 | 4 - 5 | 2086 | 83 | ||
| XM_011518182.1 | XP_011516484.1 | 4 - 5 | 582 | 83 | ||
| XM_011518184.1 | XP_011516486.1 | 3 - 4 | 1145 | 83 | ||
| GenBank mRNA | AK296343.1 | - | 4 - 5 | 504 | 83 | |
| AK298820.1 | - | 3 - 4 | 335 | 83 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map