Assay Details
Gene Symbol: PQLC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099777.3 | NP_001093247.1 | 1 - 2 | 160 | 110 | |
NM_001130001.2 | NP_001123473.1 | 1 - 2 | 160 | 110 | ||
NM_001130002.2 | NP_001123474.1 | 1 - 2 | 160 | 110 | ||
NM_001243729.1 | NP_001230658.1 | 1 - 2 | 160 | 110 | ||
NR_024016.2 | 1 - 2 | 160 | 110 | |||
XM_017005754.1 | XP_016861243.1 | 1 - 2 | 114 | 110 | ||
GenBank mRNA | AA131625.1 | - | 1 - 2 | 136 | 110 | |
BC010062.2 | - | 1 - 2 | 60 | 110 | ||
BC029129.1 | - | 1 - 2 | 107 | 110 | ||
BC126187.1 | - | 1 - 2 | 127 | 8991903 | 110 | |
BC126189.1 | - | 1 - 2 | 127 | 8991905 | 110 | |
BC144066.1 | - | 1 - 2 | 127 | 110 | ||
BE905784.1 | - | 1 - 2 | 160 | 110 | ||
BX113701.1 | - | 1 - 2 | 131 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC126187.1, BC126189.1, NM_001099777.3, NM_001130001.2, NM_001130002.2, NM_001243729.1, XM_017005754.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
