Assay Details
| Gene Symbol: TPD52L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243894.1 | NP_001230823.1 | 3 - 4 | 451 | 71 | |
| GenBank mRNA | DA689198.1 | - | 3 - 4 | 350 | 71 | |
More Information
Set Membership: |
|

TPD52L2
Genomic Map