Assay Details
Gene Symbol: PSMD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261400.2 | NP_001248329.1 | 3 - 4 | 454 | 84 | |
NM_002813.6 | NP_002804.2 | 5 - 6 | 769 | 84 | ||
NR_048555.2 | 4 - 5 | 557 | 84 | |||
GenBank mRNA | AK055339.1 | - | 1 - 2 | 1725 | 84 | |
BC002383.2 | - | 5 - 6 | 729 | 2961610 | 84 | |
BC004184.2 | - | 5 - 6 | 729 | 2961610 | 84 | |
BC004213.2 | - | 5 - 6 | 729 | 2961610 | 84 | |
BM850893.1 | - | 4 - 5 | 478 | 84 | ||
CB111716.1 | - | 3 - 4 | 399 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
