Assay Details
| Gene Symbol: YY1AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198899.1 | NP_001185828.1 | 1 - 2 | 419 | 82 | |
| NM_001198900.1 | NP_001185829.1 | 3 - 4 | 352 | 82 | ||
| NM_001198901.1 | NP_001185830.1 | 3 - 4 | 242 | 82 | ||
| NM_001198902.1 | NP_001185831.1 | 3 - 4 | 209 | 82 | ||
| NM_001198903.1 | NP_001185832.1 | 3 - 4 | 729 | 82 | ||
| NM_001198904.1 | NP_001185833.1 | 3 - 4 | 729 | 82 | ||
| NM_001198905.1 | NP_001185834.1 | 4 - 5 | 393 | 82 | ||
| NM_001198906.1 | NP_001185835.1 | 4 - 5 | 377 | 82 | ||
| NM_018253.3 | NP_060723.2 | 5 - 6 | 474 | 82 | ||
| NM_139118.2 | NP_620829.1 | 4 - 5 | 377 | 82 | ||
| NM_139119.2 | NP_620830.1 | 4 - 5 | 390 | 82 | ||
| GenBank mRNA | AF203474.1 | - | 3 - 4 | 409 | 82 | |
| AF206328.1 | - | 4 - 5 | 309 | 82 | ||
| AF466401.1 | - | 3 - 4 | 183 | 82 | ||
| AK023976.1 | - | 4 - 5 | 345 | 82 | ||
| AK297562.1 | - | 3 - 4 | 542 | 82 | ||
| AK298904.1 | - | 4 - 5 | 373 | 82 | ||
| AK303046.1 | - | 3 - 4 | 392 | 82 | ||
| AK303386.1 | - | 1 - 2 | 397 | 82 | ||
| AK309701.1 | - | 3 - 4 | 548 | 82 | ||
| AK315049.1 | - | 4 - 5 | 424 | 82 | ||
| AY604179.1 | - | 3 - 4 | 214 | 82 | ||
| BC001655.2 | - | 5 - 6 | 404 | 2961648 | 82 | |
| BC001843.2 | - | 5 - 6 | 404 | 2961648 | 82 | |
| BC008766.2 | - | 3 - 4 | 286 | 3051391 | 82 | |
| BC014906.2 | - | 4 - 5 | 322 | 3922542 | 82 | |
| BC044887.1 | - | 3 - 4 | 349 | 5276742 | 82 | |
| BG325066.1 | - | 3 - 4 | 210 | 82 | ||
| CR749789.1 | - | 4 - 5 | 364 | 82 | ||
| DC300265.1 | - | 3 - 4 | 242 | 82 | ||
| DC346197.1 | - | 1 - 2 | 419 | 82 | ||
| DC388975.1 | - | 4 - 5 | 378 | 82 | ||
| DC399389.1 | - | 4 - 5 | 393 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF206328.1 transcripts.
Set Membership: |
|

YY1AP1
Genomic Map