Assay Details
| Gene Symbol: HYPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199885.1 | NP_001186814.1 | 1 - 2 | 359 | 79 | |
| GenBank mRNA | AK022435.1 | - | 1 - 2 | 181 | 79 | |
| BQ478474.1 | - | 1 - 2 | 157 | 79 | ||
More Information
Set Membership: |
|

HYPK
Genomic Map