Assay Details
| Gene Symbol: ZNF107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282359.1 | NP_001269288.1 | 1 - 2 | 201 | 69 | |
| NM_001282360.1 | NP_001269289.1 | 1 - 2 | 201 | 69 | ||
| GenBank mRNA | DA723595.1 | - | 1 - 2 | 201 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001282359.1, NM_001282360.1 transcripts.
Set Membership: |
|

ZNF107
Genomic Map