Assay Details
Gene Symbol: ZNF561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152289.2 | NP_689502.2 | 5 - 6 | 523 | 100 | |
XM_005260150.2 | XP_005260207.1 | 6 - 7 | 661 | 100 | ||
XM_005260151.2 | XP_005260208.1 | 4 - 5 | 364 | 100 | ||
XM_006722938.2 | XP_006723001.1 | 5 - 6 | 478 | 100 | ||
XM_011528433.2 | XP_011526735.1 | 5 - 6 | 742 | 100 | ||
XM_011528434.2 | XP_011526736.1 | 6 - 7 | 648 | 100 | ||
XM_011528436.2 | XP_011526738.1 | 6 - 7 | 662 | 100 | ||
XM_011528437.2 | XP_011526739.1 | 6 - 7 | 653 | 100 | ||
XM_017027478.1 | XP_016882967.1 | 6 - 7 | 657 | 100 | ||
XM_017027479.1 | XP_016882968.1 | 5 - 6 | 546 | 100 | ||
XM_017027480.1 | XP_016882969.1 | 4 - 5 | 402 | 100 | ||
XM_017027481.1 | XP_016882970.1 | 4 - 5 | 595 | 100 | ||
GenBank mRNA | AK122974.1 | - | 4 - 5 | 402 | 100 | |
AK293004.1 | - | 5 - 6 | 513 | 100 | ||
BC012144.2 | - | 4 - 5 | 444 | 100 | ||
BC032668.1 | - | 5 - 6 | 515 | 5540009 | 100 | |
BP230901.1 | - | 5 - 6 | 523 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005260151.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
