Assay Details
| Gene Symbol: CDK11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291345.1 | NP_001278274.1 | 17 - 18 | 2086 | 86 | |
| NM_001787.2 | NP_001778.2 | 17 - 18 | 2155 | 86 | ||
| NM_033486.2 | NP_277021.2 | 17 - 18 | 2116 | 86 | ||
| NM_033487.2 | NP_277022.1 | 15 - 16 | 1856 | 86 | ||
| NM_033489.2 | NP_277024.2 | 18 - 19 | 2163 | 86 | ||
| NM_033490.2 | NP_277025.1 | 16 - 17 | 1979 | 86 | ||
| XM_006711065.3 | XP_006711128.1 | 17 - 18 | 2867 | 86 | ||
| XM_011542490.2 | XP_011540792.1 | 17 - 18 | 2924 | 86 | ||
| XM_011542491.2 | XP_011540793.1 | 17 - 18 | 2380 | 86 | ||
| XM_011542492.2 | XP_011540794.1 | 17 - 18 | 2231 | 86 | ||
| XM_011542493.2 | XP_011540795.1 | 17 - 18 | 2894 | 86 | ||
| XM_011542494.2 | XP_011540796.1 | 18 - 19 | 2971 | 86 | ||
| XM_011542495.2 | XP_011540797.1 | 18 - 19 | 2941 | 86 | ||
| XM_011542496.2 | XP_011540798.1 | 12 - 13 | 1411 | 86 | ||
| XM_017002925.1 | XP_016858414.1 | 17 - 18 | 2885 | 86 | ||
| XM_017002926.1 | XP_016858415.1 | 17 - 18 | 2855 | 86 | ||
| XM_017002927.1 | XP_016858416.1 | 17 - 18 | 2847 | 86 | ||
| XM_017002928.1 | XP_016858417.1 | 18 - 19 | 2944 | 86 | ||
| GenBank mRNA | AF067512.1 | - | 17 - 18 | 2030 | 86 | |
| AF067513.1 | - | 15 - 16 | 1770 | 86 | ||
| AF067514.1 | - | 18 - 19 | 2044 | 86 | ||
| AF067515.1 | - | 17 - 18 | 2069 | 86 | ||
| AF067516.1 | - | 18 - 19 | 2077 | 86 | ||
| AF067517.1 | - | 16 - 17 | 1893 | 86 | ||
| AF174497.1 | - | 11 - 12 | 1269 | 86 | ||
| AK000081.1 | - | 16 - 17 | 1914 | 86 | ||
| AK304679.1 | - | 17 - 18 | 1994 | 86 | ||
| BC139777.1 | - | 11 - 12 | 1199 | 86 | ||
| BC140714.1 | - | 17 - 18 | 1989 | 9021582 | 86 | |
| BC171773.1 | - | 17 - 18 | 1989 | 9054427 | 86 | |
| U04816.1 | - | 17 - 18 | 2015 | 86 | ||
| U04817.1 | - | 17 - 18 | 1988 | 86 | ||
| U04818.1 | - | 16 - 17 | 1884 | 86 | ||
| U04824.1 | - | 17 - 18 | 2021 | 86 | ||
More Information
Set Membership: |
|

CDK11B
Genomic Map