Assay Details
Gene Symbol: SNAI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178310.3 | NP_840101.1 | 2 - 3 | 762 | 75 | |
GenBank mRNA | BC041461.1 | - | 2 - 3 | 669 | 75 | |
BC107058.2 | - | 2 - 3 | 762 | 40005947 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
